Merck
  • Pharmacological induction of selective endoplasmic reticulum retention as a strategy for cancer therapy.
Pharmacological induction of selective endoplasmic reticulum retention as a strategy for cancer therapy.
Nature communications (2020-03-13)
Mohamed Mahameed, Shatha Boukeileh, Akram Obiedat, Odai Darawshi, Priya Dipta, Amit Rimon, Gordon McLennan, Rosi Fassler, Dana Reichmann, Rotem Karni, Christian Preisinger, Thomas Wilhelm, Michael Huber, Boaz Tirosh
ABSTRACT
The integrated stress response (ISR) converges on eIF2α phosphorylation to regulate protein synthesis. ISR is activated by several stress conditions, including endoplasmic reticulum (ER) stress, executed by protein kinase R-like endoplasmic reticulum kinase (PERK). We report that ER stress combined with ISR inhibition causes an impaired maturation of several tyrosine kinase receptors (RTKs), consistent with a partial block of their trafficking from the ER to the Golgi. Other proteins mature or are secreted normally, indicating selective retention in the ER (sERr). sERr is relieved upon protein synthesis attenuation and is accompanied by the generation of large mixed disulfide bonded complexes, including ERp44. sERr was pharmacologically recapitulated by combining the HIV-protease inhibitor nelfinavir with ISRIB, an experimental drug that inhibits ISR. Nelfinavir/ISRIB combination is highly effective to inhibit the growth of RTK-addicted cell lines and hepatocellular (HCC) cells in vitro and in vivo. Thus, pharmacological sERr can be utilized as a modality for cancer treatment.
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Anti-Human Serum antibody produced in rabbit, fractionated antiserum, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), peroxidase conjugate, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Dasatinib
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
Lopinavir, ≥98% (HPLC)
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Sigma-Aldrich
AMG PERK 44, ≥98% (HPLC)