Merck
  • Region specific knock-out reveals distinct roles of chromatin modifiers in adult neurogenic niches.
Region specific knock-out reveals distinct roles of chromatin modifiers in adult neurogenic niches.
Cell cycle (Georgetown, Tex.) (2018-02-13)
Christopher T Rhodes, Giulia Zunino, Shu-Wei Angela Huang, Sandra M Cardona, Astrid E Cardona, Mitchel S Berger, Vance P Lemmon, Chin-Hsing Annie Lin
ABSTRACT
Histone methyltransferases (HMTs) are present in heterogeneous cell populations within the adult brain including neurogenic niches. Yet the question remains whether loss of HMTs and the resulting changes in histone methylation alter cell fate in a region-specific manner. We utilized stereotaxic injection of Cre recombinant protein into the adult neurogenic niches, the subventricular zone (SVZ) adjacent to the lateral ventricle and the subgranular zone (SGZ) of the dentate gyrus. We confirmed that Cre protein was enzymatically active in vivo and recombination events were restricted to the vicinity of injection areas. In this study, we focus on using Cre mediated recombination in mice harboring floxed HMT: enhancer of zeste homolog 2 (EZH2) or suppressor of variegation homolog (Suv4-20h). Injectable Cre protein successfully knocked out either EZH2 or Suv4-20h, allowing assessment of long-term effects in a region-specific fashion. We performed meso-scale imaging and flow cytometry for phenotype analysis and unbiased quantification. We demonstrated that regional loss of EZH2 affects the differentiation paradigm of neural stem progenitor cells as well as the maintenance of stem cell population. We further demonstrated that regional loss of Suv4-20h influences the cell cycle but does not affect stem cell differentiation patterns. Therefore, Cre protein mediated knock-out a given HMT unravel their distinguishable and important roles in adult neurogenic niches. This Cre protein-based approach offers tightly-controlled knockouts in multiple cell types simultaneously for studying diverse regulatory mechanisms and is optimal for region-specific manipulation within complex, heterogeneous brain architectures.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-DCX Antibody, clone 2G5, clone 2G5, from mouse
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse