Merck
  • Characterization of physicochemical properties and antioxidant activity of polysaccharides from shoot residues of bamboo (Chimonobambusa quadrangularis): Effect of drying procedures.
Characterization of physicochemical properties and antioxidant activity of polysaccharides from shoot residues of bamboo (Chimonobambusa quadrangularis): Effect of drying procedures.
Food chemistry (2019-05-06)
Guangjing Chen, Changfeng Li, Shasha Wang, Xiaofei Mei, Hongxin Zhang, Jianquan Kan
ABSTRACT
In the present study, the bamboo shoot (Chimonobambusa quadrangularis) residue (BSR)-derived polysaccharides (CPS) were dried using different drying procedures, including hot air-, vacuum-, freeze-, and spray-drying. The influences of different drying procedures on the chemical compositions, physicochemical characteristics, and antioxidant activity of CPS were investigated. The results indicate that freeze-dried CPS possessed the highest polysaccharide yield (8.86%) and uronic acid content (9.42%), and the lowest medium-high molecular weight (117.49 kDa) and glucose content (5.81 mol%). Also, freeze-dried CPS had better water (5.17 g/g) and oil retention capacities (2.32 g/g) than hot air- and vacuum-dried CPS. Moreover, freeze-dried CPS exhibited the most potent antioxidant ability. Based on correlation analysis, a higher uronic acid content, lower molecular weight and lower glucose content might be closely associated with the better antioxidant activity of the freeze-dried CPS. Hence, the above results revealed freeze-drying was the optimal method for industrial production of high quality CPS.
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
Sigma-Aldrich
3-Phenylphenol, 85%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Phenazine, 98%
Sigma-Aldrich
Phenazine methosulfate
Supelco
D-(+)-Galacturonic acid monohydrate, analytical standard
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG