Skip to Content
Merck
All Photos(3)

Key Documents

262250

Sigma-Aldrich

3-Phenylphenol

85%

Synonym(s):

3-Hydroxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4OH
CAS Number:
Molecular Weight:
170.21
Beilstein:
1907938
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

85%

mp

75-80 °C (lit.)

functional group

phenyl

SMILES string

Oc1cccc(c1)-c2ccccc2

InChI

1S/C12H10O/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9,13H

InChI key

UBXYXCRCOKCZIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3 -Phenylphenol is a sensitive colorimetric reagent, used for the analysis of uronic acid.

Other Notes

remainder 4-phenylphenol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Szewczyk et al.
Molecules (Basel, Switzerland), 23(3) (2018-03-15)
Preliminary characterization and bioactivity of water-soluble polysaccharides from four Impatiens species-I. glandulifera Royle, I. parviflora DC., I. balsamina L., and I. noli-tangere L.-were investigated. The yields of polysaccharides range widely from 1.97% for I. parviflora roots to 18.63% for I.
Colorimetric determination of hexuronic acids in plant materials.
Scott RW.
Analytical Chemistry, 51(7), 936-941 (1979)
Cristóbal Narváez et al.
Chemosphere, 144, 775-784 (2015-09-29)
Effects of pesticides on non-target organisms have been studied in several taxa at different levels of biological organization, from enzymatic to behavioral responses. Although the physiological responses may be associated with higher energy costs, little is known about metabolic costs
Sue Jung Lee et al.
Journal of ginseng research, 44(4), 570-579 (2020-07-04)
Many researchers reported that the various immune activities of red ginseng are due to acid polysaccharides. But, the exact structural characteristics of the acidic polysaccharide in red ginseng have not been fully elucidated. Therefore, we isolated the acidic polysaccharide from
F K Higson et al.
Applied and environmental microbiology, 55(4), 946-952 (1989-04-01)
Isolates able to grow on 3- or 4-hydroxybiphenyl (HB) as the sole carbon source were obtained by enrichment culture. The 3-HB degrader Pseudomonas sp. strain FH12 used an NADPH-dependent monooxygenase restricted to 3- and 3,3'-HBs to introduce an ortho-hydroxyl. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service