Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cryo-Babies
®
Labels
Products
Product Category
labels (7)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Length (in)
Color
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z366218
Cryo-Babies
®
Labels,
white, L × W 1.28 in. × 0.5 in., roll of 1000 labels
Expand
View Pricing
Z743334
Cryo-Babies
®
Labels,
green, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing
Z743333
Cryo-Babies
®
Labels,
blue, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing
Z743337
Cryo-Babies
®
Labels,
white, L × W 0.94 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing
Z743335
Cryo-Babies
®
Labels,
orange, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing
Z743336
Cryo-Babies
®
Labels,
red, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing
Z743338
Cryo-Babies
®
Labels,
yellow, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Expand
View Pricing