
2,2,3,3-Tetrafluoro-1,4-butanediol

Synonym(s):
2,2,3,3-Tetrafluorobutanediol, NSC 95113, Tetrafluoro-1,4-butanediol
Linear Formula:
HOCH2CF2CF2CH2OH
CAS Number:
Molecular Weight:
162.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1