Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
scopolamine, hyoscine
manufacturer/tradename
USP
mp
195-199 °C (dry matter) (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4
InChI
1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1
InChI key
LACQPOBCQQPVIT-SEYKEWMNSA-N
Gene Information
human ... CHRM1(1128)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Belladonna Extract
- Belladonna Extract Tablets
- Belladonna Leaf
- Belladonna Tincture
- Homatropine Hydrobromide
- Methscopolamine Bromide
- Methscopolamine Bromide Tablets
Biochem/physiol Actions
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description meets USP testing specifications | description Pharmaceutical Secondary Standard; Certified Reference Material | description phyproof® Reference Substance |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename PhytoLab |
| grade pharmaceutical primary standard | grade - | grade certified reference material, pharmaceutical secondary standard | grade primary reference standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) food and beverages |
| format neat | format - | format neat | format neat |
| API family scopolamine, hyoscine | API family - | API family scopolamine, hyoscine | API family - |
| mp 195-199 °C (dry matter) (lit.) | mp 195-199 °C (dry matter) (lit.) | mp - | mp - |
Still not finding the right product?
Explore all of our products under Scopolamine hydrobromide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



