Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H32O7
CAS Number:
Molecular Weight:
432.51
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
grade
pharmaceutical primary standard
agency
USP
API family
schisandrin
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
COc1cc2C[C@H](C)[C@@](C)(O)Cc3cc(OC)c(OC)c(OC)c3-c2c(OC)c1OC
InChI
1S/C24H32O7/c1-13-9-14-10-16(26-3)20(28-5)22(30-7)18(14)19-15(12-24(13,2)25)11-17(27-4)21(29-6)23(19)31-8/h10-11,13,25H,9,12H2,1-8H3/t13-,24-/m0/s1
InChI key
YEFOAORQXAOVJQ-RZFZLAGVSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Schisandrin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Northern Schisandra Fruit
- Northern Schisandra Fruit Dry Extract
- Northern Schisandra Fruit Powder
Biochem/physiol Actions
Antioxidant; Anti-inflammatory, reduces oxidative stress.
Schizandrin is a natural product with several biological properties involved with antioxidant and anti-inflammatory activities. It reduces the formation of ROS, inhibits the mitochondrial pathway of the apoptotic process and oxidative stress. Schizandrin has been reported to have hepatoprotective, antitumor, antiviral, and antiamnesic effects, and has neuroprotective activity against glutamate induced neurotoxicity.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.