Skip to Content
Merck

Skip To

1598019

USP

Rabeprazole sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-([4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl)-1H-benzo[d]imidazole, Pariprazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H20N3O3S · Na
CAS Number:
Molecular Weight:
381.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

rabeprazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

[Na+].COCCCOc1ccnc(CS(=O)c2nc3ccccc3[n-]2)c1C

InChI

1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1

InChI key

KRCQSTCYZUOBHN-UHFFFAOYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rabeprazole sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Rabeprazole sodium is gastric proton pump inhibitor. It suppresses the production of acid in the stomach by inhibiting the gastric H+/K+ATPase (hydrogen-potassium adenosine triphosphatase) at the secretory surface of the gastric parietal cell. Rabeprazole sodium has been used clinically to treat acid-reflux disorders (GERD), peptic ulcer disease, H. pylori eradication, and prevent gastroinetestinal bleeds associated with NSAID use.
Rabeprazole sodium is gastric proton pump inhibitor; antiulcerative.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1525Y0001778SML0476
description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

description

≥98% (HPLC)

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

format

neat

format

neat

format

neat

format

-

agency

USP

agency

traceable to USP 1598019

agency

-

agency

-

Gene Information

human ... ATP4A(495), ATP4B(496)

Gene Information

human ... ATP4A(495), ATP4B(496)

Gene Information

human ... ATP4A(495), ATP4B(496)

Gene Information

human ... ATP4A(495), ATP4B(496)


Still not finding the right product?

Explore all of our products under Rabeprazole sodium


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters