Skip to Content
Merck

Skip To

1478593

USP

Ondansetron Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-[(Dimethylamino)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride, 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H20N2O · HCl
CAS Number:
Molecular Weight:
292.80
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
11612579
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

ondansetron

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)CC1CCc2c(C1=O)c3ccccc3n2C

InChI

1S/C16H20N2O.ClH/c1-17(2)10-11-8-9-14-15(16(11)19)12-6-4-5-7-13(12)18(14)3;/h4-7,11H,8-10H2,1-3H3;1H

InChI key

GFQLILQFQHMEPF-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ondansetron Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Ondansetron Oral Solution
  • Ondansetron Injection
  • Ondansetron
  • Ondansetron Hydrochloride
  • Ondansetron Orally Disintegrating Tablets
  • Ondansetron Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2223PHR2224PHR2221
description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

Pharmaceutical Secondary Standard; Certified Reference Material

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

API family

ondansetron

API family

ondansetron

API family

ondansetron

API family

ondansetron

agency

USP

agency

traceable to USP 1478593

agency

traceable to USP 1478618

agency

traceable to Ph. Eur. Y0000195, traceable to USP 1478629


Still not finding the right product?

Explore all of our products under Ondansetron Related Compound A


pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Met. Corr. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters