Skip to Content
Merck

1443500

USP

Miconazole nitrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-Miconazole nitrate salt, 1-(2,4-Dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl)imidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

miconazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC1=CC(Cl)=CC=C1C(OCC2=CC=C(Cl)C=C2Cl)CN3C=CN=C3.[O-][N+](O)=O

InChI

1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)

InChI key

MCCACAIVAXEFAL-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Miconazole nitrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Miconazole Nitrate Cream
  • Miconazole Nitrate Topical Powder
  • Miconazole Nitrate Vaginal Suppositories

Biochem/physiol Actions

Antifungal azole. Mode of action: Inhibits cytochrome P450-dependent 14α-demethylase, which is critical to ergosterol biosynthesis. The accumulated 14α-methylated sterols change the membrane structure of sensitive fungi, resulting in an altered cell membrane permeability. Also inhibits peroxidases, which results in accumulation of peroxide within the cell.
Miconazole interacts with 14-α demethylase, a cytochrome P-450 enzyme necessary for ergosterol biosynthesis. The inhibition of ergosterol results in increased cellular permeability causing leakage of cellular contents. Miconazole may also inhibit endogenous respiration, interact with phospholipids in the cell membrane, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and interfere with triglyceride and phospholipid biosynthesis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Miconazole nitrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Stanislaw A Buechner
The Journal of dermatological treatment, 25(3), 226-231 (2013-04-06)
This study investigated the non-inferiority of efficacy and tolerance of 2% miconazole nitrate shampoo in comparison with 2% ketoconazole shampoo in the treatment of scalp seborrheic dermatitis. A randomized, double-blind, comparative, parallel group, multicenter study was done. A total of
S Gebremedhin et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 65(4), 593-600 (2014-09-03)
Oral candidiasis in the form of Candida-associated denture stomatitis (CaDS) is associated with Candida adhesion and biofilm formation on the fitting surface of poly (methyl methacrylate) (PMMA) dentures. Candida biofilms show considerable resistance to most conventional antifungal agents, a phenomenon
Bart Pauwels et al.
The journal of sexual medicine, 11(7), 1664-1674 (2014-05-21)
Erectile dysfunction (ED), as well as many cardiovascular diseases, is associated with impaired nitric oxide (NO) bioavailability. Recently, oxime derivatives have emerged as vasodilators due to their NO-donating capacities. However, whether these oximes offer therapeutic perspectives as an alternative NO



Global Trade Item Number

SKUGTIN
1443500-200MG04061838735577