1375003
USP
Maltol
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
3-Hydroxy-2-methyl-4-pyrone, 3-Hydroxy-2-methyl-4H-pyran-4-one, Larixinic acid, Maltol
About This Item
grade
pharmaceutical primary standard
API family
maltol
autoignition temp.
1364 °F
expl. lim.
25 %
manufacturer/tradename
USP
mp
160-164 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC1=C(O)C(=O)C=CO1
InChI
1S/C6H6O3/c1-4-6(8)5(7)2-3-9-4/h2-3,8H,1H3
InChI key
XPCTZQVDEJYUGT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Ethyl Maltol
- Metoprolol Tartrate and Hydrochlorothiazide Tablets
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service