Skip to Content
Merck
All Photos(1)

Documents

1359404

USP

Levetiracetam

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, 2(S)-(2-Oxopyrrolidin-1-yl)butyramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levetiracetam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC[C@H](N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1

InChI key

HPHUVLMMVZITSG-LURJTMIESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levetiracetam USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Levetiracetam is a pyrrolidine with antiepileptic activity. Stereoselective binding of levetiracetam was confined to synaptic plasma membranes in the central nervous system with no binding occurring in peripheral tissue. Levetiracetam inhibits burst firing without affecting normal neuronal excitability, which suggests that it may selectively prevent hyper-synchronization of epileptiform burst firing and propagation of seizure activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad U Farooq et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 66(6), 541-561 (2009-03-07)
The role of levetiracetam in different epileptic, nonepileptic, neurologic, and psychiatric disorders is discussed. Levetiracetam, an antiepileptic drug (AED), was first approved as an adjunctive therapy for the treatment of partial epilepsy in adults. It is currently being used in
Rafal M Kaminski et al.
Epilepsia, 50(3), 387-397 (2008-07-17)
Levetiracetam (Keppra) is an antiepileptic drug (AED) characterized by a novel mechanism of action, unique profile of activity in seizure models, and broad-spectrum clinical efficacy. The present report critically reviews several preclinical studies focused on combination therapy with levetiracetam and
J Rösche et al.
Fortschritte der Neurologie-Psychiatrie, 81(1), 21-27 (2012-08-15)
Non-convulsive status epilepticus and epilepsia partialis continua are common epileptic conditions for which straightforward recommendations based on controlled randomised trials for treatment of therapy refractory courses are lacking. Therefore in these conditions sometimes antiepileptic drugs that are not approved by
Justin B Usery et al.
Journal of neuro-oncology, 99(2), 251-260 (2010-02-11)
To determine the safety and tolerability of IV and oral levetiracetam monotherapy for seizures in brain tumor patients following resection. Brain tumor patients undergoing neurosurgery with >or=1 seizure within the preceding month prior to surgery were enrolled to receive intravenous
Katherine A Lyseng-Williamson
Drugs, 71(4), 489-514 (2011-03-15)
Levetiracetam (Keppra®, E Keppra®) is an established second-generation antiepileptic drug (AED). Worldwide, levetiracetam is most commonly approved as adjunctive treatment of partial onset seizures with or without secondary generalization; other approved indications include monotherapy treatment of partial onset seizures with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service