Skip to Content
Merck

1353113

USP

Diluted Isosorbide Mononitrate Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,4:3,6-Dianhydro-D-glucitol 2-nitrate in lactose, Isosorbide-2-mononitrate in lactose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

isosorbide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diluted Isosorbide Mononitrate Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diluted Isosorbide Mononitrate
  • Isosorbide Dinitrate Extended-Release Tablets
  • Isosorbide Mononitrate Extended-Release Tablets
  • Isosorbide Mononitrate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library