Skip to Content
Merck
All Photos(1)

Documents

1295607

USP

Glycerol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

product name

Glycerin, United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

API family

glycerin

autoignition temp.

698 °F

manufacturer/tradename

USP

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glycerin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Glycerin Ophthalmic Solution
  • Glyceryl Dibehenate
  • Mono- and Di-glycerides
  • Ethylcellulose Dispersion Type B
  • L-Alpha-Glycerylphosphorylcholine

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glyceryl Dibehenate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(4), 5791-5791 (2018)
Glycerin
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 28(4), 2132-2132 (2020)
L-alpha-Glycerylphosphorylcholine
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 47(3) (2021)
Mono- and Di-glycerides
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(4), 5890-5890 (2018)
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service