Skip to Content
Merck

1270720

USP

Flavoxate Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 3-methyl-4-oxo-2-phenyl-4H-benzopyrane-8-carboxylate, 3-Methylflavone-8-carboxylic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H14O4
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

flavoxate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[o]1c2c([c](c(c1c3ccccc3)C)=O)cccc2C(=O)OC

InChI

1S/C18H14O4/c1-11-15(19)13-9-6-10-14(18(20)21-2)17(13)22-16(11)12-7-4-3-5-8-12/h3-10H,1-2H3

InChI key

OSHFQLSUZICPRA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Flavoxate Related Compound B


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library