Skip to Content
Merck

1242010

USP

Erythromycin B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

12-Deoxyerythromycin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H67NO12
CAS Number:
Molecular Weight:
717.93
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

erythromycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N(C1C(C(OC(C1)C)O[C@H]2[C@@](C[C@H](C(=O)[C@@H]([C@H]([C@H]([C@H](OC(=O)[C@@H]([C@H](C2C)OC3OC(C(C(C3)(OC)C)O)C)C)CC)C)O)C)C)(O)C)O)(C)C

InChI

1S/C37H67NO12/c1-14-26-20(4)29(40)21(5)28(39)18(2)16-36(9,44)33(50-35-30(41)25(38(11)12)15-19(3)46-35)22(6)31(23(7)34(43)48-26)49-27-17-37(10,45-13)32(42)24(8)47-27/h18-27,29-33,35,40-42,44H,14-17H2,1-13H3/t18-,19?,20+,21+,22?,23-,24?,25?,26-,27?,29+,30?,31+,32?,33-,35?,36-,37?/m1/s1

InChI key

IDRYSCOQVVUBIJ-ZTOAHLHUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Erythromycin is an antibiotic produced by growth of certain strains of Streptomyces erythreus. This product is composed largely of erythromycin A with small amounts of erythromycins B and C and is recommended for concentration at 100 mg/L. Concentrations between 50 and 200 mg/L have also proven effective in controlling bacterial growth. Erythromycin has been used as a motilin receptor agonist, to block respiratory glycoconjugate secretion in human airways in vitro, and for selecting plasmid-cured and recombinant lactococcus lactis MG1363 strains.

Biochem/physiol Actions

Mode of Action: Erythromycin acts by inhibiting elongation at the transpeptidation step, specifically aminoacyl translocation from the A-site to P-site.

Antimicrobial Spectrum: This product acts against both gram-negative and gram-positive bacteria.

Preparation Note

This product is soluble in water at 2 mg/mL, with a 0.067% solution in water yielding a pH of 8.0-10.5. It is also soluble in ethanol at 50 mg/mL, yielding a clear, colorless to faint yellow solution. It is freely soluble in alcohol, acetone, chloroform, acetonitrile and ethyl acetate but forms salts with acids. All solutions should be protected from light.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Legal Information

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Disclaimer

This product is stable in solution at 37°C for 3 days. Stock solutions should be stored at 2-8°C.


Still not finding the right product?

Explore all of our products under Erythromycin B


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library