Skip to Content
Merck

Skip To

1225281

USP

Dorzolamide hydrochloride

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H16N2O4S3 · HCl
CAS Number:
Molecular Weight:
360.90
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5896026
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dorzolamide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(N)(=O)=O

InChI

1S/C10H16N2O4S3.ClH/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16;/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16);1H/t6-,8-;/m0./s1

InChI key

OSRUSFPMRGDLAG-QMGYSKNISA-N

Gene Information

human ... CA2(760)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dorzolamide hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dorzolamide Hydrochloride and Timolol Maleate Ophthalmic Solution
  • Dorzolamide Hydrochloride Ophthalmic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP1069Y0000726PHR1925
description

United States Pharmacopeia (USP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

EDQM

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

Gene Information

human ... CA2(760)

Gene Information

human ... CA2(760)

Gene Information

human ... CA2(760)

Gene Information

human ... CA2(760)

agency

USP

agency

-

agency

-

agency

traceable to Ph. Eur. Y0000726, traceable to USP 1225281


Still not finding the right product?

Explore all of our products under Dorzolamide hydrochloride


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

target_organs

Central nervous system,Gastrointestinal tract,Bone,Blood,Bladder

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters