Skip to Content
Merck

Skip To

1222818

USP

Dissolution Performance Verification Standard - Prednisone (30 tablets)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

DPVS-Prednisone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

prednisone

form

tablet

manufacturer/tradename

USP

concentration

10 mg/tablet (nominal prednisone content per tablet)

application(s)

pharmaceutical

format

mixture

storage temp.

room temp

General description

The USP Performance Verification Test (PVT) is an essential aspect of the General Chapter <711> Dissolution that assesses the comprehensive performance of Dissolution Apparatus 1 (basket) and Dissolution Apparatus 2 (paddle). DPVS - Prednisone Tablets is specially engineered by USP to use as a reference standard for PVT of apparatus used in dissolution testing for the following reasons:

  • High stability and ease of use
  • High sensitivity to operational and mechanical variables of instrument setup
  • Low sensitivity to media degassing
  • Low tablet-to-tablet variability
These features help enable you to identify any differences from the ideal dissolution instrument setup so you can have confidence in your dissolution testing results.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Find answers to frequently asked questions on USP products in USP FAQs.

Application

Dissolution Performance Verification Standard (DPVS) – Prednisone tablets USP reference standard is used for the comprehensive qualification of dissolution instruments using performance verification testing (PVT) to achieve reliable results that reflect the quality of the drug product.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Each tablet contains: Dicalcium Phosphate Anhydrous (7757-93-9), Microcrystalline Cellulose (9004-34-6), Colloidal silicone dioxide (112945-52-5), Magnesium Stearate (557-04-0), Prednisone (53-03-2), Sodium Dodecyl Sulfate Fine (151-21-3), Sodium Starch Glycolate Type A (9063-38-1)
Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
155900615580031557000
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

mixture

format

neat

format

neat

format

neat

form

tablet

form

-

form

-

form

-

concentration

10 mg/tablet (nominal prednisone content per tablet)

concentration

-

concentration

-

concentration

-


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2 Inhalation

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters