Skip to Content
Merck
All Photos(1)

Documents

1158504

USP

Cyclosporine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cyclosporin A, Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cyclosporine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclosporine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cyclosporine Capsules
  • Cyclosporine Injection
  • Cyclosporine Oral Solution
  • Cyclosporine Compounded Ophthalmic Solution, Veterinary

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cyclosporine
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(4) (2021)
Cyclosporine Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(1), 1216-1216 (2013)
Cyclosporine Compounded Ophthalmic Solution, Veterinary
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 1217-1217 (2020)
Cyclosporine Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(1), 1214-1214 (2018)
Cyclosporine Capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(1), 1213-1213 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service