Skip to Content
Merck

1154900

USP

Cyclomethicone 6

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dodecamethylcyclohexasiloxane, 2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H36O6Si6
CAS Number:
Molecular Weight:
444.92
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

cyclomethicone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C

InChI

1S/C12H36O6Si6/c1-19(2)13-20(3,4)15-22(7,8)17-24(11,12)18-23(9,10)16-21(5,6)14-19/h1-12H3

InChI key

IUMSDRXLFWAGNT-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclomethicone 6 USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Cyclomethicone

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


wgk

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

signalword

Danger

hcodes

Hazard Classifications

PBT - vPvB

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library