Skip to Content
Merck

Skip To

1154569

USP

Beta Cyclodextrin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

β-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
78623
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant

grade

pharmaceutical primary standard

agency

USP

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

analyte chemical class(es)

oligosaccharides

manufacturer/tradename

USP

mp

290-300 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

General description

Betadex is a nonreducing cyclic compound composed of seven alpha-(1–4) linked d-glucopyranosyl units.
Find answers to frequently asked questions on USP products in USP FAQs.

Application

Beta Cyclodextrin USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare standard and system suitability solution for the assay and impurity analysis by using liquid chromatography coupled with Refractive index detector according to the below monographs of United States Pharmacopeia (USP) :

  • Betadex
  • Alfadex

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1439239760C4767
application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

-

application(s)

-

format

neat

format

neat

format

-

format

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

biological source

plant

biological source

-

biological source

-

biological source

plant


Still not finding the right product?

Explore all of our products under Beta Cyclodextrin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is it suitable for GC and LCMS?

    1 answer
    1. Although this product is not tested specifically for GC or LCMS , it is suitable for liquid chromatography applications in accordance to USP.

      Helpful?

Reviews

No rating value

Active Filters