Skip to Content
Merck

1111103

USP

Chlorhexidine acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Chlorhexidine diacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H30Cl2N10 · 2C2H4O2
CAS Number:
Molecular Weight:
625.55
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

chlorhexidine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)N\C(=N\C(=N)NCCCCCCNC(=N)\N=C(\Nc2ccc(cc2)Cl)/N)\N.OC(=O)C.OC(=O)C

InChI

1S/C22H30Cl2N10.2C2H4O2/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*1-2(3)4/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H3,(H,3,4)

InChI key

WDRFFJWBUDTUCA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorhexidine acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Chlorhexidine Acetate Topical Solution
  • Chlorhexidine Gluconate Oral Rinse
  • Chlorhexidine Gluconate Solution
  • Chlorhexidine Gluconate Topical Gel
  • Chlorhexidine Gluconate Topical Solution
  • Chlorhexidine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Chlorhexidine acetate


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 2

target_organs

Liver,Teeth

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Wen-Shuai Liu et al.
Chemical biology & drug design, 85(1), 91-97 (2014-09-13)
Five N-methyl-N-R-N,N-bis(2-hydroxyethyl) ammonium bromides (R = -benzyl (chloride, BNQAS), -dodecyl (C12QAS), -tetradecyl (C14QAS), -hexadecyl (C16QAS), -octadecyl (C18QAS)) were prepared based on N-methyldiethanolamine (MDEA) and halohydrocarbon. Five QAS were characterized by FTIR, NMR, and MS. BNQAS, C12QAS, C14QAS, and C16QAS were
Manikandan Ekambaram et al.
Journal of dentistry, 42(7), 872-882 (2014-02-25)
To evaluate the effect of adjunctive application of ethanol-wet bonding and chlorhexidine (CHX) with a hydrophobic adhesive on bond durability of fibre posts to intraradicular dentine. Ninety-six extracted human teeth with a single root and root canal were prepared for
Manikandan Ekambaram et al.
Journal of dentistry, 42(6), 709-719 (2014-04-16)
To examine the effect of adjunctive application of chlorhexidine (CHX) and ethanol-wet bonding (EWB) on bond durability and nanoleakage of hydrophobic adhesive to sound (SD) and caries-affected dentine (CAD). Dentine surfaces of molars were etched after caries removal and randomly



Global Trade Item Number

SKUGTIN
1111103-200MG04061838698520