Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
API family
celecoxib
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Cc1ccc(cc1)-c2cc(nn2-c3ccc(cc3)S(N)(=O)=O)C(F)(F)F
InChI
1S/C17H14F3N3O2S/c1-11-2-4-12(5-3-11)15-10-16(17(18,19)20)22-23(15)13-6-8-14(9-7-13)26(21,24)25/h2-10H,1H3,(H2,21,24,25)
InChI key
RZEKVGVHFLEQIL-UHFFFAOYSA-N
Gene Information
human ... PTGS2(5743)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Biochem/physiol Actions
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename BP | manufacturer/tradename - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| API family celecoxib | API family celecoxib | API family celecoxib | API family celecoxib |
| Gene Information human ... PTGS2(5743) | Gene Information human ... PTGS2(5743) | Gene Information human ... PTGS2(5743) | Gene Information human ... PTGS2(5743) |
Still not finding the right product?
Explore all of our products under Celecoxib
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 1 - Repr. 1B - STOT RE 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1098504-200MG | 04061838693150 |


