Skip to Content
Merck
All Photos(1)

Key Documents

1096779

USP

Casticin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3′,5-Dihydroxy-3,4′,6,7-tetramethoxyflavone, 3,4′,6,7-Tetra-O-methylquercetagetin, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4H-1-benzopyran-4-one, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one, Quercetagetin 3,4′,6,7-tetramethyl ether, Vitexicarpin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
Beilstein:
1300392
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

casticin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=C(OC)C(=O)c3c(O)c(OC)c(OC)cc3O2

InChI

1S/C19H18O8/c1-23-11-6-5-9(7-10(11)20)17-19(26-4)16(22)14-12(27-17)8-13(24-2)18(25-3)15(14)21/h5-8,20-21H,1-4H3

InChI key

PJQLSMYMOKWUJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Casticin is a flavonoid, widely used as an anti-inflammatory agent. It has anti-tumor properties and is considered to be a potent immunomodulatory and cytotoxic compound.

Application

It was may be used as reference standard to compare retention time and ion peaks in plant extracts during GC/MS analysis to quantify Casticin and artemetin.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ahmed Mesaik et al.
Phytotherapy research : PTR, 23(11), 1516-1520 (2009-05-15)
Casticin (1), a flavonoid isolated from the aerial parts of Vitex agnus-castus, was found to be a potent immunomodulatory and cytotoxic compound. The activity was tested in vitro for chemiluminescence, chemotaxis, T-cell proliferation and cytotoxicity. Casticin (1) exhibited a significant
Pamela J Weathers et al.
Planta medica, 78(10), 1024-1026 (2012-06-08)
A number of flavonoids including casticin and artemetin from Artemisia annua have shown synergism with artemisinin against Plasmodium falciparum, but it is unclear if the flavonoids are also extracted into a tea infusion of the plant. Using a tea infusion
Jia-Kun Shen et al.
Annals of hematology, 88(8), 743-752 (2009-01-14)
Casticin, a component from Vitex rotundifolia, widely used as an anti-inflammatory agent in Chinese traditional medicine, was reported to have anti-tumor activities. This study aims to examine the anti-leukemic activity of casticin on leukemia cells and its molecular mechanism. Cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service