Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3956
grade
pharmaceutical primary standard
agency
USP
vapor pressure
400 mmHg ( 323 °C)
API family
carprofen
manufacturer/tradename
USP
bp
355 °C (lit.)
mp
243-246 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
c1ccc2c(c1)[nH]c3ccccc23
InChI
1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H
InChI key
UJOBWOGCFQCDNV-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Carprofen Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Carprofen
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Carprofen Related Compound A
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 4 - Carc. 2 - Muta. 2
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
