Skip to Content
Merck

Skip To

1096407

USP

Carboplatin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

cis-Diammine(1,1-cyclobutanedicarboxylato) platinum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
[C4H6(CO2)2]Pt(NH3)2
CAS Number:
Molecular Weight:
371.25
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

carboplatin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N.N.O=C1O[Pt]OC(=O)C12CCC2

InChI

1S/C6H8O4.2H3N.Pt/c7-4(8)6(5(9)10)2-1-3-6;;;/h1-3H2,(H,7,8)(H,9,10);2*1H3;/q;;;+2/p-2

InChI key

OLESAACUTLOWQZ-UHFFFAOYSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carboplatin USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, diluted standard, sensitivity, and system suitability solutions for the assay, and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Carboplatin
  • Carboplatin Injection
  • Carboplatin for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP711C2538PHR3417
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

API family

carboplatin

API family

carboplatin

API family

-

API family

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Carboplatin


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters