Skip to Content
Merck

Skip To

1071304

USP

Bile salts

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Bile salts are amphipathic molecules derived from cholesterol in the liver. They are incorporated into selective media to inhibit the growth of non-enteric organisms, thereby facilitating the isolation and enumeration of enteric pathogens.

not a USP categoty

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Bile salts USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pancreatin
  • Pancreatin Tablets
  • Pancrelipase
  • Pancrelipase Capsules
  • Pancrelipase Delayed-Release Capsules
  • Pancrelipase Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Bile salts USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
101354513910001287722
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

form

powder

form

-

form

-

form

powder

format

neat

format

neat

format

neat

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

agency

USP

agency

USP

agency

USP

agency

USP


Still not finding the right product?

Explore all of our products under Bile salts


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters