Select a Size
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
azithromycin
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
N1([C@@H]([C@H]([C@@]([C@H](OC(=O)[C@@H]([C@H]([C@@H]([C@H]([C@@](C[C@H](C1)C)(O)C)O[C@@H]3O[C@@H](C[C@@H]([C@H]3O)NC)C)C)O[C@@H]2O[C@H]([C@@H]([C@@](C2)(OC)C)O)C)C)CC)(O)C)O)C)C
InChI
1S/C37H70N2O12/c1-14-26-37(10,45)30(41)23(6)39(12)18-19(2)16-35(8,44)32(51-34-28(40)25(38-11)15-20(3)47-34)21(4)29(22(5)33(43)49-26)50-27-17-36(9,46-13)31(42)24(7)48-27/h19-32,34,38,40-42,44-45H,14-18H2,1-13H3/t19-,20-,21+,22-,23-,24+,25+,26-,27+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
InChI key
FZYUKBLYECHAGN-HOQMJRDDSA-N
General description
Application
Also used to prepare standard stock, system suitability, standard, system suitability stock, and peak identification solution for impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
- Azithromycin for Injection
- Azithromycin
- Azithromycin Tablets
Analysis Note
Other Notes
Still not finding the right product?
Explore all of our products under N-Demethylazithromycin
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1046089-15MG | 04061833849200 |