Skip to Content
Merck

Skip To

1019803

USP

Aminobenzoic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Aminobenzoic acid, PABA, Vitamin Bx, Vitamin H1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
471605
eCl@ss:
32160406
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

aminobenzoic acid

manufacturer/tradename

USP

mp

187-189 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aminobenzoic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Aminobenzoic Acid Gel
  • Antipyrine and Benzocaine Otic Solution
  • Benzocaine
  • Benzocaine and Menthol Topical Aerosol®
  • Benzocaine Cream
  • Benzocaine Gel

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Aerosol is a registered trademark of Cytec Technology Corp.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR184001973A9878
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

peptide synthesis

API family

aminobenzoic acid

API family

aminobenzoic acid

API family

-

API family

-

mp

187-189 °C (lit.)

mp

187-189 °C (lit.)

mp

186-188 °C, 187-189 °C (lit.)

mp

187-189 °C (lit.)


Still not finding the right product?

Explore all of our products under Aminobenzoic acid


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

339.8 °F - closed cup

flash_point_c

171 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters