Key Documents
Z227331
Frit for Supelco® Filter
stainless steel, pore size 2.0 μm, pkg of 5 ea
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
material
stainless steel
packaging
pkg of 5 ea
pore size
2.0 μm
Application
This product may be used to protect HPLC columns containing 5 μm or larger particles against particulate matter that gets accumulated during the chromatographic operations.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.