PHL89825
Wogonin
phyproof® Reference Substance
Synonym(s):
5,7-Dihydroxy 8-methoxyflavone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
SMILES string
O1c2c(c(cc(c2OC)O)O)C(=O)C=C1c3ccccc3
InChI
1S/C16H12O5/c1-20-15-12(19)7-10(17)14-11(18)8-13(21-16(14)15)9-5-3-2-4-6-9/h2-8,17,19H,1H3
InChI key
XLTFNNCXVBYBSX-UHFFFAOYSA-N
Related Categories
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Plabon K Das et al.
Anti-cancer agents in medicinal chemistry, 19(15), 1796-1808 (2019-07-06)
Cancer Stem Cells (CSCs) are the subpopulation of cancer cells which are directly involved in drug resistance, metastases to distant organ and cancer recurrence. A systematic literature search was conducted through various electronic databases including, Pubmed, Scopus, Google scholar using
Yong Wang et al.
Frontiers in pharmacology, 11, 1152-1152 (2020-08-15)
Wogonin, an active component derived from Scutellaria baicalensis, has shown anti-tumor activities in several malignancies. However, the roles of wogonin in RCC cells remain elusive. Here, we explored the effects of wogonin on RCC cells and the underlying mechanisms. We
Ying Liang et al.
Frontiers in genetics, 11, 701-701 (2020-07-28)
Andrographis paniculata (Chinese name: Chuanxinlian) is an annual dicotyledonous medicinal plant widely grown in China and Southeast Asia. The dried plant has a highly acclaimed usage in the traditional Chinese medicine for its antipyretic, anti-inflammatory, and analgesic effects. In order
Wogonin glucuronidation in liver and intestinal microsomes of humans, monkeys, dogs, rats, and mice.
Nobumitsu Hanioka et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(8), 906-912 (2020-02-02)
Wogonin, one of the flavonoids isolated from Scutellaria baicalensis, exhibits some beneficial bioactivities, including anti-inflammatory and anticancer effects, and is metabolized into glucuronide by UDP-glucuronosyltransferase (UGT) enzymes in humans. In the present study, wogonin glucuronidation was examined in the liver
Li Fan et al.
Journal of immunology research, 2020, 3032425-3032425 (2020-06-23)
Wogonin (5,7-dihydroxy-8-methoxyflavone) is an ingredient of the extracts from Scutellaria baicalensis, which has documented a wide spectrum of anti-inflammatory and antitumor activities, including inhibiting regulatory T cells, regulating effector T cell functions, and mediating macrophage immunity. However, the potential effect
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service