Skip to Content
Merck
All Photos(1)

Key Documents

PHL89379

Limonin

phyproof® Reference Substance

Synonym(s):

Citrolimonin, Dictamnolactone, Evodin, Limonoic acid 3,19:16,17 dilactone, Obaculactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30O8
CAS Number:
Molecular Weight:
470.51
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

CC1(C)O[C@H]2CC(=O)OC[C@]23[C@H]4CC[C@@]5(C)[C@@H](OC(=O)[C@H]6O[C@@]56[C@]4(C)C(=O)C[C@@H]13)c7ccoc7

InChI

1S/C26H30O8/c1-22(2)15-9-16(27)24(4)14(25(15)12-31-18(28)10-17(25)33-22)5-7-23(3)19(13-6-8-30-11-13)32-21(29)20-26(23,24)34-20/h6,8,11,14-15,17,19-20H,5,7,9-10,12H2,1-4H3/t14-,15-,17-,19-,20+,23-,24-,25+,26+/m0/s1

InChI key

KBDSLGBFQAGHBE-MSGMIQHVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Limonin may be used in assays to quantify limonin in citrus products. It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.

Biochem/physiol Actions

Limonin is a triterpenoid aglycone that is a bitter principle of citrus fruits. It has anti-proliferative, proapoptotic activity on several cancer cell lines and inhibits azoxymethane-induced colon cancer in rats. It also inhibits HIV-1 replication in culturedf monocytes, macrophages, and mononuclear cells, perhaps by inhibition of HIV-1 protease activity.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service