G001719
HPLC Analysis of Carboxymethylated Apohemoglobin (Horse) Tryptic Digest on Discovery® BIO Wide Pore C18
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Discovery BIO Wide Pore C18, 15 cm × 2.1 mm I.D., 3 μm particles (568222-U)
mobile phase: [A] 0.1% TFA in water; [B] 0.1% TFA in water:0.1% TFA in acetonitrile, 50:50;
gradient: 0-100% B in 50 min
flow rate: 0.2 mL/min
column temp.: 35°C
detector: UV, 220 nm
injection: 5 μL, each at 0.8 mg/mL in 0.1% TFA
suitability
application for HPLC
application(s)
life science and biopharma
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service