Skip to Content
Merck
All Photos(1)

Key Documents

CRM06783

Supelco

Chlorfluazuron

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H9Cl3F5N3O3
CAS Number:
Molecular Weight:
540.65
Beilstein:
8369967
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2cc(Cl)c(Oc3ncc(cc3Cl)C(F)(F)F)c(Cl)c2

InChI

1S/C20H9Cl3F5N3O3/c21-10-5-9(30-19(33)31-17(32)15-13(24)2-1-3-14(15)25)6-11(22)16(10)34-18-12(23)4-8(7-29-18)20(26,27)28/h1-7H,(H2,30,31,32,33)

InChI key

UISUNVFOGSJSKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Chlorfluazuron belongs to the family of benzoylurea insecticides used against various crops, fruits, and vegetables. It is a chitin synthesis inhibitor and controls the lepidopteran pests by disrupting molting and subsequent completion of their life cycle due to incomplete exoskeleton formation.
As per the commission regulation no 1107/2009, repealing the council directive 91/414/EEC, chlorfluazuron is not approved for use in the European Union. But according to Art 18(b) Reg 396/2005, a default maximum residue limit (MRL) of 0.01 mg/kg is applicable, wherever necessary.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Chlorfluazuron may also be used as follows:

  • To study the metabolism of chlorfluazuron and flonicamid in tea samples using a modified QuEChERS extraction and UPLC-MS/MS quantification
  • Determination of chlorfluazuron residues in Chinese cabbage, Chinese kale, Chinese lettuce, and cauliflower by ultra high-performance liquid chromatography-tandem mass spectrometry(UHPLC-MS/MS)
  • Development and validation of a gas chromatography-mass spectrometry (GC-MS) method for the detection and measurement of chlorfluazuron in termites
  • To determine 9 benzoylurea insecticides in samples of whole milk, rice, egg, lemon, lettuce, beef meat, and avocado using pressurized liquid extraction and liquid chromatography-mass spectrometry(LC-MS)
  • To develop a method based on in-syringe dispersive liquid-liquid microextraction and HPLC determination for the analysis of 5 benzoylurea insecticides in water and tea beverage samples
  • Analysis of benzoylurea insecticides in tea samples using Fe3O4- hyperbranched polyester (HBPE) nanocomposite as an adsorbent for magnetic solid phase extraction (MSPE) and HPLC

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingna Sun et al.
International journal of environmental research and public health, 16(10) (2019-05-22)
Chlorfluazuron is used as a highly effective insect growth regulator to control a variety of crop pests. However, residues of this pesticide have been shown to be harmful to human health. To evaluate the residual dissipation pattern and risk for
Hongxia Li et al.
The Science of the total environment, 754, 142070-142070 (2020-09-14)
Degradation and metabolism of chlorfluazuron and flonicamid from tea garden to cup were simultaneously investigated by a modified QuEChERS method coupled with UPLC-MS/MS quantification. The dissipation half-lives of chlorfluazuron, flonicamid, and total flonicamid (the sum of flonicamid and its metabolites
A simple and sensitive assay using GC-MS for determination of chlorfluazuron in termites
Lee C, et al.
Journal of Pesticide Science, D13- 036 (2013)
Magnetic solid-phase extraction of benzoylurea insecticides in tea samples with Fe3O4-hyperbranched polyester magnetic composite as sorbent
Liu C, et al.
Journal of Separation Science, 42(8), 1610- 1619 (2019)
Huazi Wang et al.
Talanta, 162, 625-633 (2016-11-14)
A novel in-syringe dispersive liquid-liquid microextraction based on the solidification of ionic liquids (in-syringe SIL-DLLME) was coupled with high-performance liquid chromatography-ultraviolet detector (HPLC-UVD) to detect five benzoylurea insecticides (BUs) in water and tea beverage samples. In this method, the hydrophobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service