BP1118
1-Methylpiperazine
British Pharmacopoeia (BP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
vapor density
3.5 (vs air)
API family
cyclizine
form
crystalline powder
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
refractive index
n20/D 1.466 (lit.)
bp
138 °C (lit.)
density
0.903 g/mL at 25 °C (lit.)
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
−20°C
SMILES string
CN1CCNCC1
InChI
1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3
InChI key
PVOAHINGSUIXLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Also used in monographs such as:
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
88.7 °F - closed cup
Flash Point(C)
31.5 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service