Skip to Content
Merck
All Photos(1)

Key Documents

96690

Supelco

Isobutyl vinyl ether

analytical standard

Synonym(s):

Vinyl isobutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OCH=CH2
CAS Number:
Molecular Weight:
100.16
Beilstein:
1560159
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

grade

analytical standard

Quality Level

vapor density

3.45 (vs air)

vapor pressure

68 mmHg ( 20 °C)

Assay

≥95.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.395 (lit.)

bp

82-83 °C (lit.)

mp

−112 °C (lit.)

density

0.768 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)COC=C

InChI

1S/C6H12O/c1-4-7-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

OZCMOJQQLBXBKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service