Skip to Content
Merck
All Photos(1)

Key Documents

94043

Supelco

Isoxaflutole Metabolite RPA202248

PESTANAL®, analytical standard

Synonym(s):

α-(Cyclopropylcarbonyl)-2-(methylsulfonyl)-β-oxo-4-(trifluoromethyl)benzenepropanenitrile, (2RS)-3-Cyclopropyl-2-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]-3-oxopropanenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12F3NO4S
CAS Number:
Molecular Weight:
359.32
MDL number:

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
liquid chromatography (LC): suitable

application(s)

agriculture
environmental
food and beverages

format

neat

SMILES string

N#CC(C(C1CC1)=O)C(C2=CC=C(C(F)(F)F)C=C2S(=O)(C)=O)=O

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service