Skip to Content
Merck
All Photos(1)

Key Documents

91644

Supelco

2,4-Diamino-6-phenyl-1,3,5-triazine

analytical standard

Synonym(s):

6-Phenyl-1,3,5-triazine-2,4-diamine, Benzoguanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N5
CAS Number:
Molecular Weight:
187.20
Beilstein:
153223
MDL number:

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

219-221 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

Nc1nc(N)nc(n1)-c2ccccc2

InChI

1S/C9H9N5/c10-8-12-7(13-9(11)14-8)6-4-2-1-3-5-6/h1-5H,(H4,10,11,12,13,14)

InChI key

GZVHEAJQGPRDLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service