Skip to Content
Merck
All Photos(1)

Key Documents

822857

TSKgel® SuperSW mAb HR (4 µm) HPLC Column

L x I.D. 4 cm × 6 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
L × I.D.:
4 cm × 6 mm
particle size:
4 μm
matrix active group:
diol phase
pore size:
250 Å pore size
matrix:
silica particle platform

Product Name

TSKgel® SuperSW mAb HR Guard Column, phase diol, L × I.D. 4 cm × 6 mm, 4 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh SuperSW HR Guard, 22857

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1 mL/min flow rate
10-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

4 μm

pore size

250 Å pore size

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The TSKgel SuperSW and UltraSW mAb columns consist of three specialized columns designed for the separation and analysis of monoclonal antibodies (mAb). While mAbs can be analyzed using many different modes of HPLC, size exclusion is best to determine the level of aggregation, dimer formation, and fragmentation, making it the best method for heterogeneity studies. TSKgel SuperSW mAb HR columns are optimized for high resolution analysis of mAb monomer and dimer; these 30 cm x 7.8 mm ID slica-based stainless steel columns offer reduced lot-to-lot variation, long column life, reduction of unspecified adsorption, and superior recovery of aggregates. The TSKgel SuperSW mAb columns utilize a unique pore-controlled technology, which produces a shallow calibration curve in the molecular weight region of a typical monoclonal antibody. The calibration curve for the TSKgel SuperSW mAb HR column is similar to that of the industry-leading TSKgel G3000SWxl column and has a shallower slope than the TSKgel UltraSW Aggregate column around the molecular weight range of gamma-globulin. This shallow calibration curve produces high resolution separations.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service