Skip to Content
Merck

Skip To

808024

TSKgel® G6000PWXL HPLC Column

phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32110501
UNSPSC Code:
41115700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

suitable for USP L38, suitable for USP L39

manufacturer/tradename

Tosoh 08024

parameter

≤0.5 M salt concentration, ≤20% organic solvent, 1 mL/min flow rate, 20 bar max. pressure, 80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

13 μm

pore size

>1000 Å pore diameter

operating pH

2-12

separation technique

size exclusion (SEC)

storage temp.

2-8°C

General description

Use TSKgel® G6000PWXL columns for analysis of water-soluble linear polymers with molecular weights up to 8 million Da. The smaller particle size of PWXL-type columns provide 1.7 times higher resolution than their PW counterpart, making PWXL-type columns more suitable for analytical purposes.

Physical form

Shipped in water.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
805765805109808023
L × I.D.

30 cm × 7.8 mm

L × I.D.

30 cm × 7.5 mm

L × I.D.

60 cm × 7.5 mm

L × I.D.

30 cm × 7.8 mm

separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

matrix

polymer particle platform

matrix

polymer particle platform

matrix

polymer particle platform

matrix

polymer particle platform

particle size

13 μm

particle size

17 μm

particle size

17 μm

particle size

10 μm

matrix active group

hydroxylated methacrylate phase

matrix active group

hydroxylated methacrylate phase

matrix active group

hydroxylated methacrylate phase

matrix active group

hydroxylated methacrylate phase

pore size

>1000 Å pore diameter

pore size

>1000 Å pore diameter

pore size

>1000 Å

pore size

1000 Å pore diameter




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters