Skip to Content
Merck

72849

Dimethyl adipate

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3OCO(CH2)4COOCH3
CAS Number:
Molecular Weight:
174.19
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1707443
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95%

autoignition temp.

680 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.1 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.428 (lit.)

bp

109-110 °C/14 mmHg (lit.)

mp

8 °C (lit.)

density

1.062 g/mL at 20 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

COC(=O)CCCCC(=O)OC

InChI

1S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3

InChI key

UDSFAEKRVUSQDD-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Dimethyl adipate


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library