Skip to Content
Merck
All Photos(1)

Documents

58979

Supelco

SUPELCOSIL LC-CN (3 µm) HPLC Columns

L × I.D. 3.3 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-CN HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 4.6 mm

Agency

suitable for USP L10

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

4% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

3.3 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.5 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

cyano phase

particle size

3 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The LC-CN phase is often used as a substitute for silica because it offers the advantages of a bonded phase (such as quick equilibration, and less sensitivity to small changes of the water content in the mobile phase). More often, however, the LC-CN column is operated under reversed-phase mobile phase conditions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Paczkowski et al.
Polish journal of pharmacology, 47(5), 429-434 (1995-09-01)
This paper describes a simple high-performance liquid chromatographic (HPLC) method for the determination of diltiazem and desacetyldiltiazem in human serum. After basic methyl-tert-butyl ether extraction and back-extraction with hydrochloric acid, the drug and its metabolite was injected into a Supelcosil
S Fogli et al.
Therapeutic drug monitoring, 21(3), 367-375 (1999-06-12)
A single high-performance liquid chromatography (HPLC) method, suitable for the analysis of daunorubicin, idarubicin, doxorubicin, epirubicin, and their 13-dihydro metabolites is validated in the present study. Preparation of plasma samples was performed by a first extraction of analytes with a
H Kataoka et al.
Journal of analytical toxicology, 24(4), 257-265 (2000-06-29)
A simple and rapid method for the determination of amphetamine, methamphetamine, and their 3,4-methylenedioxy derivatives in urine samples was developed using automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS). In-tube SPME is an extraction technique
H Kataoka et al.
Journal of chromatography. B, Biomedical sciences and applications, 731(2), 353-359 (1999-10-08)
The technique of automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS) was evaluated for the determination of ranitidine. In-tube SPME is an extraction technique for organic compounds in aqueous samples, in which analytes are extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service