The full scale range of this gauge is 0 - 30 mm Hg. Low vacuums below 5 mm Hg are readable. Pressure graduations are marked at 0.5 mm Hg intervals.
57161-U
SPE Manifold Gauge/Bleed Valve
Remote In-Line Design
Synonym(s):
SPE Manifold Gauge/Bleed Valve
About This Item
Recommended Products
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Looking to assess our vacuum pump for lab work, can this yield small absolute pressures (<5mmHg) as with a standard Mercury U-tube manometer?
1 answer-
Helpful?
-
-
Hello, I am looking for an inline manifold valve with gauge that will fit 3/8" inner diameter pvc tubing for our vacuum manifold. Can product # 57161-U accommodate this size tubing? Thank you
1 answer-
This product will accommodate 3/16" - 1/4" tubing. 3/8" tubing will be too wide.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service