Skip to Content
Merck
All Photos(2)

Documents

55276-U

Supelco

HybridSPE®-Phospholipid solid phase extraction (SPE) Cartridge

Cartridge, bed wt. 30 mg, volume 1 mL, pk of 200, polypropylene material (hardware), PE frit (20 μm)

Synonym(s):

HybridSPE (phospholipid and protein removal) SPE cartridge tube, 6 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

HybridSPE®-Phospholipid, Cartridge, bed wt. 30 mg, volume 1 mL, pk of 200, polypropylene material (hardware), PE frit (20 μm)

material

PE frit (20 μm)
polypropylene hardware

Quality Level

composition

bed wt., 30 mg

packaging

pk of 200

technique(s)

solid phase extraction (SPE): suitable

volume

1 mL

matrix active group

zirconia-based phase

Looking for similar products? Visit Product Comparison Guide

General description

HybridSPE-Phospholipid technology is a simple and generic sample prep platform designed for the gross level removal of endogenous protein and phospholipid interferences from biological plasma and serum prior to LC-MS or LC-MS/MS analysis. Biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is loaded on the HybridSPE-Phospholipid 96-well plate or cartridge which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE-Phospholipid stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC-MS or LC-MS-MS analysis.

The "In-well" and "In-cartridge" precipitation methods are available for the HybridSPE-Phospholipid 96-well version and HybridSPE-Phospholipid Ultra cartridge in which biological plasma/serum is first added to either the well or cartridge, followed by acidified acetonitrile (precipitation agent). After a brief mixing/vortexing step, vacuum is applied. Because the 96-well and Ultra cartridge versions contain a series of low porosity hydrophobic filters/frits, the packed-bed filter/frit assembly acts as a depth filter facilitating the concurrent removal of both phospholipids and precipitated proteins during the extraction process. Standard HybridSPE-Phospholipid cartridges require an "off-line" precipitation method.

Application


  • Less is more: a methodological assessment of extraction techniques for per- and polyfluoroalkyl substances (PFAS) analysis in mammalian tissues.: This study evaluates various extraction techniques, including the use of HybridSPE-Phospholipid, for analyzing PFAS in mammalian tissues. The research highlights the importance of effective phospholipid removal to ensure accurate PFAS quantification (Mertens et al., 2023).

  • Rapid analysis of 65 pharmaceuticals and 7 personal care products in plasma and whole-body tissue samples of fish using acidic extraction, zirconia-coated silica cleanup, and liquid chromatography-tandem mass spectrometry.: The study includes HybridSPE-Phospholipid as part of the cleanup process, demonstrating its efficiency in removing phospholipids to improve the detection of pharmaceuticals and personal care products in biological samples (Tanoue et al., 2020).

  • Multi LC-MS/MS and LC-HRMS Methods for Determination of 24 Mycotoxins including Major Phase I and II Biomarker Metabolites in Biological Matrices from Pigs and Broiler Chickens.: This research utilizes HybridSPE-Phospholipid for the cleanup of biological matrices, enhancing the accuracy of mycotoxin detection by effectively removing interfering phospholipids (Lauwers et al., 2019).

  • Effective phospholipid removal from plasma samples by solid phase extraction with the use of copper (II) modified silica gel cartridges.: The study compares various phospholipid removal techniques, highlighting the performance of HybridSPE-Phospholipid in achieving high phospholipid removal efficiency from plasma samples (Flieger et al., 2017).

  • Liquid chromatography mass spectrometry determination of perfluoroalkyl acids in environmental solid extracts after phospholipid removal and on-line turbulent flow chromatography purification.: The research demonstrates the application of HybridSPE-Phospholipid in the removal of phospholipids from environmental solid extracts, facilitating the accurate measurement of perfluoroalkyl acids (Mazzoni et al., 2016).

Features and Benefits

  • Merges the simplicity of protein precipitation and the selectivity of SPE via the targeted removal of phospholipids
  • Reduce ion-suppression through the complete removal of phospholipids and precipitated proteins
  • 2-3 step generic procedure
  • Minimal to no method development
  • Available in 96-well and 1 mL cartridge dimensions

Legal Information

HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of an HPLC-MS/MS method to determine clopidogrel in human plasma. Use of incurred samples to test back-conversion
Silvestro, L., et al.
Journal of Chromatography. B, Biomedical Applications, 878 (30), 3134-3142 (2010)
Solid-phase extraction of phosphorous-containing amino acid herbicides from biological specimens with a zirconia-coated silica cartridge
Watanabe, D., et al.
Journal of Chromatography. B, Biomedical Applications, 969, 69-76 (2014)
Quantitative determination of capecitabine and its six metabolites in human plasma using liquid chromatography coupled to electrospray tandem mass spectrometry
Deenen, M, et al.
Journal of Chromatography. B, Biomedical Applications, 913-914, 30-40 (2013)
C Ferreiro-Vera et al.
Journal of chromatography. A, 1240, 21-28 (2012-04-17)
Fractionation of biological fluids for proper analysis by LC-MS of low-abundance metabolites in the presence of high-abundant endogenous metabolites is of interest, particularly when the latter cause undesirable phenomena such as ionization suppression, as is the case with phospholipids (PLs).
Nora Unceta et al.
Journal of pharmaceutical and biomedical analysis, 70, 529-533 (2012-06-01)
This work proposes a liquid chromatography-electrospray ionization ion trap mass spectrometry (LC-ESI-ITMS) method, for the quantification of sildenafil (SDF), tadalafil (TDF) and vardenafil (VDF) and their metabolites N-desmethylSDF, O-desethylSDF and N-desethylVDF, preceded by a sample preparation step based on protein

Articles

An article focusing on ion-suppression and phospholipid contamination and some of their major causes and difficulties.

An article focusing on ion-suppression and phospholipid contamination and some of their major causes and difficulties.

An article focusing on ion-suppression and phospholipid contamination and some of their major causes and difficulties.

An article focusing on ion-suppression and phospholipid contamination and some of their major causes and difficulties.

Protocols

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service