Skip to Content
Merck
All Photos(1)

Documents

54794

Supelco

LiChrospher® RP-18 (5 µm) HPLC Columns

L × I.D. 1 cm × 4.6 mm, HPLC guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

LiChrospher® RP-18 HPLC Guard Cartridge, 5 μm particle size, L × I.D. 1 cm × 4.6 mm

Agency

suitable for USP L1

Quality Level

feature

endcapped: no

packaging

pkg of 4 ea

extent of labeling

21% Carbon loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix

Octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

compatibility

for use with 3.0 to 4.6 mm columns

separation technique

reversed phase

General description

Use guard cartridge holder 54987

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service