Skip to Content
Merck
All Photos(1)

Documents

54039-U

Supelco

SPE tube

10%AGNO3/44%H2SO4/SI, volume 12mL

Synonym(s):

12mL 10% silver nitrate/44% sulfuric acid SPE tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

12mL 10%AGNO3/44%H2SO4/SI SPE Tube, pk of 20 ea

packaging

pk of 20 ea

Quality Level

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

storage temp.

2-8°C

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service