509450
SGE Gas Tight Syringes, Luer Lock
50F-LL-GT, volume 50 μL, needle size (not included)
Synonym(s):
SYRINGE SGE 50UL, LUER LOCK
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
glass syringe
needle size
(not included)
feature
Fixed Luer Lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 004230
volume
50 μL
General description
SGE gas tight syringe without a needle, Model: 50F-LL-GT (volume 50 μL)
- Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
- Autoclavable syringes but the plunger should be removed.
- Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
- Maximum inertness for liquid or gas samples.
- Suitable for manual dispenser/diluter applications.
- High accuracy of dispensed volume.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)