Skip to Content
Merck
All Photos(1)

Key Documents

50541-U

Supelco

Ascentis® Express 90 Å C18 (5 μm) HPLC Columns

L × I.D. 5 mm × 3 mm, HPLC Guard Cartridge pkg of 3 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Ascentis®

feature

endcapped

packaging

pkg of 3 ea

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

5 mm × 3 mm

surface area

90 m2/g

matrix

superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

90 Å

operating pH

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Ascentis® Express Guard Columns provide physical (filtration) and chemical protection for costly analytical columns without compromising the very high performance of Ascentis Express columns. These Ascentis Express guard columns are capable of continuous use at pressures up to 9000 psi (600 bar) with only hand-tightening. Guard cartridges are easily replaced without removing the guard column holder from the flow path. The cartridges are packed with Ascentis Express Fused-Core® articles. Order guard column holder separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

required but not provided


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Jakabová et al.
Journal of chromatography. A, 1232, 295-301 (2012-03-07)
Hyoscyamine (atropine) and scopolamine are the predominant tropane alkaloids in the Datura genus, occurring in all plant organs. The assessment of the alkaloid content of various plant parts is essential from the viewpoint of medical use, but also as a
Heena et al.
Journal of chromatographic science, 53(5), 800-806 (2014-10-18)
This study represents a new analytical high-performance liquid chromatography-fluorescence detector method for the determination of Al(III) as Al(III) complex with 8-hydroxyquinoline-5-sulfonic acid in a tap water sample and a coke sample. A micellar liquid chromatographic method is proposed for the
Petra Šilarová et al.
Food chemistry, 237, 471-480 (2017-08-03)
The degradation of catechins and other phenolics in green tea infusions were monitored using fast HPLC/MS separation. The final separation was performed within 2.5min using Ascentis Express C18 column (50mm×2.1mm i.d.) packed with 2μm porous shell particles. Degradation was studied
Judy Stone
Methods in molecular biology (Clifton, N.J.), 1378, 301-320 (2015-11-26)
Serum from bar-coded tubes, and then internal standard, are pipetted to 96-well plates with an 8-channel automated liquid handler (ALH). The first precipitation reagent (methanol:ZnSO4) is added and mixed with the 8-channel ALH. A second protein precipitating agent, 1 %
Babu Rao Chandu et al.
SpringerPlus, 2(1), 194-194 (2013-06-07)
A bioequivalence study was proved of generic Febuxostat 80 mg tablets (T) in healthy volunteers.For this purpose, Authors developed a simple, sensitive, selective, rapid, rugged and reproducible liquid chromatography-tandem mass spectrometry method for the quantification of Febuxostat (FB) in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service