Skip to Content
Merck
All Photos(1)

Documents

504815

Supelco

Supelclean LC-SAX SPE Tube

bed wt. 100 mg, volume 1 mL, pk of 108

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712

material

polypropylene tube

Quality Level

composition

bed wt., 100 mg

packaging

pk of 108

technique(s)

solid phase extraction (SPE): suitable

volume

1 mL

matrix

silica gel

matrix active group

SAX phase
quaternary amine, Cl- counterion

diameter

40-45 μm

pore size

60 Å

application(s)

food and beverages

separation technique

ion exchange
ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Supelclean LC-SAX SPE Tube is a strong anion-exchange cartridge.

Application

Supelclean LC-SAX SPE Tube may be used for the solid-phase extraction of:
  • Folic acid in infant milk formula prior to its determination by isotope dilution-liquid chromatography-tandem mass spectrometry (ID-LC-MS/MS).

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of isotope dilution-liquid chromatography/tandem mass spectrometry as a candidate reference method for the determination of folic acid in infant milk formula
Jung M, et al.
Bull. Korean Chem. Soc., 28(5), 745-745 (2007)

Articles

SPE retention mechanism in this case is based on the electrostatic attraction of charged functional groups of the analyte(s) to oppositely charged functional groups on the sorbent.

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service